Assay Details
| Gene Symbol: TRX-M4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112404.5 | NP_188155.1 | 1 - 2 | 471 | 96 | |
| GenBank mRNA | AF375443.1 | - | 1 - 2 | 300 | 96 | |
| AY060538.1 | - | 1 - 2 | 264 | 96 | ||
| AY088157.1 | - | 1 - 2 | 308 | 96 | ||
More Information
Set Membership: |
|

TRX-M4
Genomic Map