Assay Details
| Gene Symbol: AT3G15490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112417.2 | NP_188168.2 | 2 - 3 | 832 | 111 | |
| GenBank mRNA | DQ056594.1 | - | 2 - 3 | 171 | 111 | |
More Information
Set Membership: |
|

AT3G15490
Genomic Map