Assay Details
| Gene Symbol: NAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112418.4 | NP_188169.1 | 2 - 3 | 728 | 140 | |
| GenBank mRNA | AB049070.1 | - | 2 - 3 | 454 | 140 | |
| BT025659.1 | - | 2 - 3 | 454 | 140 | ||
More Information
Set Membership: |
|

NAC3
Genomic Map