Assay Details
| Gene Symbol: ROD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112452.3 | NP_566527.1 | 1 - 2 | 841 | 97 | |
| GenBank mRNA | AY086160.1 | - | 1 - 2 | 584 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROD1| Gene Symbol: ROD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112452.3 | NP_566527.1 | 1 - 2 | 841 | 97 | |
| GenBank mRNA | AY086160.1 | - | 1 - 2 | 584 | 97 | |
Set Membership: |
|