Assay Details
| Gene Symbol: SULTR3;4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112469.3 | NP_188220.1 | 2 - 3 | 461 | 105 | |
| GenBank mRNA | AB054645.1 | - | 2 - 3 | 461 | 105 | |
More Information
Set Membership: |
|

SULTR3;4
Genomic Map