Assay Details
| Gene Symbol: PDIL1-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112481.4 | NP_188232.2 | 2 - 3 | 870 | 111 | |
| GenBank mRNA | BT015349.1 | - | 2 - 3 | 692 | 111 | |
| BT015798.1 | - | 2 - 3 | 637 | 111 | ||
More Information
Set Membership: |
|

PDIL1-6
Genomic Map