Assay Details
| Gene Symbol: AT3G16160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112486.3 | NP_188237.2 | 2 - 3 | 496 | 67 | |
| GenBank mRNA | AJ630495.1 | - | 2 - 3 | 239 | 67 | |
| AY568667.1 | - | 2 - 3 | 239 | 67 | ||
More Information
Set Membership: |
|

AT3G16160
Genomic Map