Assay Details
| Gene Symbol: DELTA-TIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112495.4 | NP_188245.1 | 2 - 3 | 555 | 119 | |
| GenBank mRNA | AK229954.1 | - | 1927 | 119 | ||
| AY085921.1 | - | 2 - 3 | 453 | 119 | ||
| U39485.1 | - | 2 - 3 | 398 | 119 | ||
More Information
Set Membership: |
|

DELTA-TIP
Genomic Map