Assay Details
| Gene Symbol: ABCG29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202982.2 | NP_001189911.1 | 4 - 5 | 1023 | 99 | |
| NM_112505.4 | NP_566543.1 | 4 - 5 | 1269 | 99 | ||
More Information
Set Membership: |
|

ABCG29
Genomic Map