Assay Details
| Gene Symbol: PAB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112509.2 | NP_188259.1 | 2 - 3 | 675 | 72 | |
| GenBank mRNA | AX765882.1 | - | 2 - 3 | 542 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAB6| Gene Symbol: PAB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112509.2 | NP_188259.1 | 2 - 3 | 675 | 72 | |
| GenBank mRNA | AX765882.1 | - | 2 - 3 | 542 | 72 | |
Set Membership: |
|