Assay Details
| Gene Symbol: PAB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112509.2 | NP_188259.1 | 3 - 4 | 940 | 131 | |
| GenBank mRNA | AX765882.1 | - | 3 - 4 | 807 | 131 | |
More Information
Set Membership: |
|

PAB6
Genomic Map