Assay Details
| Gene Symbol: TPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112557.4 | NP_188306.2 | 1 - 2 | 265 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPR2| Gene Symbol: TPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112557.4 | NP_188306.2 | 1 - 2 | 265 | 153 | |
Set Membership: |
|