Assay Details
| Gene Symbol: POK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338286.1 | NP_001326050.1 | 28 - 29 | 5846 | 69 | |
| NM_001338287.1 | NP_001326049.1 | 28 - 29 | 5805 | 69 | ||
| NM_112614.4 | NP_188362.2 | 28 - 29 | 5822 | 69 | ||
| GenBank mRNA | DQ399529.1 | - | 28 - 29 | 5594 | 69 | |
More Information
Set Membership: |
|

POK1
Genomic Map