Assay Details
| Gene Symbol: GAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161149.1 | NP_001154621.1 | 2 - 3 | 206 | 86 | |
| NM_112657.2 | NP_188403.1 | 1 - 2 | 136 | 86 | ||
More Information
Set Membership: |
|

GAD5
Genomic Map