Assay Details
| Gene Symbol: PYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112662.4 | NP_188408.1 | 4 - 5 | 1117 | 115 | |
| GenBank mRNA | AF545062.1 | - | 4 - 5 | 828 | 115 | |
| AY035029.1 | - | 4 - 5 | 956 | 115 | ||
| AY059103.1 | - | 4 - 5 | 828 | 115 | ||
More Information
Set Membership: |
|

PYD1
Genomic Map