Assay Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112672.3 | NP_566592.1 | 2 - 3 | 495 | 67 | |
| GenBank mRNA | AF182953.1 | - | 2 - 3 | 351 | 67 | |
| BT024740.1 | - | 2 - 3 | 298 | 67 | ||
More Information
Set Membership: |
|

SURF1
Genomic Map