Assay Details
| Gene Symbol: DAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112674.4 | NP_566593.1 | 2 - 3 | 313 | 139 | |
| GenBank mRNA | AY039550.1 | - | 2 - 3 | 282 | 139 | |
| AY093751.1 | - | 2 - 3 | 249 | 139 | ||
More Information
Set Membership: |
|

DAC
Genomic Map