Assay Details
| Gene Symbol: XPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112681.3 | NP_188427.2 | 3 - 4 | 483 | 159 | |
| GenBank mRNA | AF197940.1 | - | 3 - 4 | 469 | 159 | |
| AF367299.1 | - | 3 - 4 | 408 | 159 | ||
| AY058175.1 | - | 3 - 4 | 483 | 159 | ||
| AY091683.1 | - | 3 - 4 | 268 | 159 | ||
More Information
Set Membership: |
|

XPL1
Genomic Map