Assay Details
| Gene Symbol: MOS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112699.3 | NP_566599.1 | 4 - 5 | 699 | 110 | |
| GenBank mRNA | AY050910.1 | - | 4 - 5 | 637 | 110 | |
| AY088112.1 | - | 4 - 5 | 620 | 110 | ||
| AY091412.1 | - | 4 - 5 | 576 | 110 | ||
| EF688334.1 | - | 4 - 5 | 576 | 110 | ||
More Information
Set Membership: |
|

MOS4
Genomic Map