Assay Details
| Gene Symbol: LPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112706.3 | NP_566602.1 | 6 - 7 | 950 | 90 | |
| GenBank mRNA | AK229475.1 | - | 6 - 7 | 888 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPP4| Gene Symbol: LPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112706.3 | NP_566602.1 | 6 - 7 | 950 | 90 | |
| GenBank mRNA | AK229475.1 | - | 6 - 7 | 888 | 90 | |
Set Membership: |
|