Assay Details
| Gene Symbol: SHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202994.1 | NP_001189923.1 | 2 - 3 | 281 | 80 | |
| NM_112723.3 | NP_188467.2 | 2 - 3 | 292 | 80 | ||
| NM_202604.3 | NP_974333.1 | 2 - 3 | 368 | 80 | ||
| GenBank mRNA | AY093042.1 | - | 2 - 3 | 281 | 80 | |
| BT003428.1 | - | 2 - 3 | 143 | 80 | ||
More Information
Set Membership: |
|

SHH2
Genomic Map