Assay Details
| Gene Symbol: EMB1865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112724.4 | NP_188468.1 | 5 - 6 | 2125 | 74 | |
| GenBank mRNA | AY063908.1 | - | 5 - 6 | 2111 | 74 | |
More Information
Set Membership: |
|

EMB1865
Genomic Map