Assay Details
| Gene Symbol: AT3G18470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112732.3 | NP_188476.1 | 1 - 2 | 184 | 62 | |
| GenBank mRNA | AY065153.1 | - | 1 - 2 | 184 | 62 | |
| AY081605.1 | - | 1 - 2 | 60 | 62 | ||
More Information
Set Membership: |
|

AT3G18470
Genomic Map