Assay Details
| Gene Symbol: AT3G18470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112732.3 | NP_188476.1 | 3 - 4 | 466 | 66 | |
| GenBank mRNA | AY065153.1 | - | 3 - 4 | 466 | 66 | |
| AY081605.1 | - | 3 - 4 | 342 | 66 | ||
More Information
Set Membership: |
|

AT3G18470
Genomic Map