Assay Details
| Gene Symbol: CASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112733.3 | NP_566611.1 | 11 - 12 | 1513 | 116 | |
| GenBank mRNA | AY056225.1 | - | 11 - 12 | 1457 | 116 | |
| AY142680.1 | - | 11 - 12 | 1281 | 116 | ||
More Information
Set Membership: |
|

CASP
Genomic Map