Assay Details
| Gene Symbol: CASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112733.3 | NP_566611.1 | 14 - 15 | 1818 | 84 | |
| GenBank mRNA | AY056225.1 | - | 14 - 15 | 1762 | 84 | |
| AY142680.1 | - | 14 - 15 | 1586 | 84 | ||
More Information
Set Membership: |
|

CASP
Genomic Map