Assay Details
| Gene Symbol: TSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112759.5 | NP_188503.2 | 17 - 18 | 3594 | 106 | |
| GenBank mRNA | AB112744.1 | - | 17 - 18 | 3552 | 106 | |
More Information
Set Membership: |
|

TSK
Genomic Map