Assay Details
| Gene Symbol: TSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112759.5 | NP_188503.2 | 3 - 4 | 1400 | 89 | |
| GenBank mRNA | AB112744.1 | - | 3 - 4 | 1358 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSK| Gene Symbol: TSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112759.5 | NP_188503.2 | 3 - 4 | 1400 | 89 | |
| GenBank mRNA | AB112744.1 | - | 3 - 4 | 1358 | 89 | |
Set Membership: |
|