Assay Details
| Gene Symbol: Tic62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112775.4 | NP_188519.2 | 10 - 11 | 1333 | 66 | |
| GenBank mRNA | BT002039.1 | - | 10 - 11 | 1290 | 66 | |
| BT008403.1 | - | 10 - 11 | 1146 | 66 | ||
More Information
Set Membership: |
|

Tic62
Genomic Map