Assay Details
| Gene Symbol: ETP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084716.2 | NP_001078185.1 | 1 | 1605 | 135 | |
| NM_112784.3 | NP_566622.1 | 1 | 1642 | 135 | ||
| GenBank mRNA | AY045839.1 | - | 1 | 1377 | 135 | |
More Information
Set Membership: |
|

ETP1
Genomic Map