Assay Details
| Gene Symbol: POK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112791.6 | NP_188535.4 | 17 - 18 | 2465 | 66 | |
| GenBank mRNA | DQ399530.1 | - | 16 - 17 | 2265 | 66 | |
More Information
Set Membership: |
|

POK2
Genomic Map