Assay Details
| Gene Symbol: POK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112791.6 | NP_188535.4 | 29 - 30 | 7562 | 79 | |
| GenBank mRNA | DQ399530.1 | - | 28 - 29 | 7362 | 79 | |
More Information
Set Membership: |
|

POK2
Genomic Map