Assay Details
| Gene Symbol: POK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112791.6 | NP_188535.4 | 7 - 8 | 1351 | 104 | |
| GenBank mRNA | DQ399530.1 | - | 6 - 7 | 1151 | 104 | |
More Information
Set Membership: |
|

POK2
Genomic Map