Assay Details
| Gene Symbol: POK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112791.6 | NP_188535.4 | 10 - 11 | 1675 | 85 | |
| GenBank mRNA | DQ399530.1 | - | 9 - 10 | 1475 | 85 | |
More Information
Set Membership: |
|

POK2
Genomic Map