Assay Details
| Gene Symbol: LARP6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112796.2 | NP_188540.1 | 4 - 5 | 892 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LARP6c| Gene Symbol: LARP6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112796.2 | NP_188540.1 | 4 - 5 | 892 | 88 | |
Set Membership: |
|