Assay Details
| Gene Symbol: LARP6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338382.1 | NP_001325674.1 | 5 - 6 | 971 | 71 | |
| NM_112796.2 | NP_188540.1 | 6 - 7 | 1052 | 71 | ||
More Information
Set Membership: |
|

LARP6c
Genomic Map