Assay Details
| Gene Symbol: LARP6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338382.1 | NP_001325674.1 | 8 - 9 | 1211 | 64 | |
| NM_112796.2 | NP_188540.1 | 9 - 10 | 1292 | 64 | ||
More Information
Set Membership: |
|

LARP6c
Genomic Map