Assay Details
| Gene Symbol: PREP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203003.1 | NP_001189932.1 | 11 - 12 | 2105 | 72 | |
| NM_112804.5 | NP_188548.2 | 11 - 12 | 2334 | 72 | ||
| GenBank mRNA | AY090240.1 | - | 11 - 12 | 2110 | 72 | |
| AY091051.1 | - | 11 - 12 | 2108 | 72 | ||
| BT002372.1 | - | 11 - 12 | 2044 | 72 | ||
| BT006362.1 | - | 11 - 12 | 2044 | 72 | ||
More Information
Set Membership: |
|

PREP1
Genomic Map