Assay Details
| Gene Symbol: PREP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203003.1 | NP_001189932.1 | 12 - 13 | 2306 | 106 | |
| NM_112804.5 | NP_188548.2 | 12 - 13 | 2535 | 106 | ||
| GenBank mRNA | AY090240.1 | - | 12 - 13 | 2311 | 106 | |
| AY091051.1 | - | 12 - 13 | 2309 | 106 | ||
| BT002372.1 | - | 12 - 13 | 2245 | 106 | ||
| BT006362.1 | - | 12 - 13 | 2245 | 106 | ||
More Information
Set Membership: |
|

PREP1
Genomic Map