Assay Details
| Gene Symbol: PREP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203003.1 | NP_001189932.1 | 1 - 2 | 379 | 65 | |
| NM_112804.5 | NP_188548.2 | 1 - 2 | 575 | 65 | ||
| GenBank mRNA | AY090240.1 | - | 1 - 2 | 351 | 65 | |
| AY091051.1 | - | 1 - 2 | 349 | 65 | ||
| BT002372.1 | - | 1 - 2 | 285 | 65 | ||
| BT006362.1 | - | 1 - 2 | 285 | 65 | ||
More Information
Set Membership: |
|

PREP1
Genomic Map