Assay Details
| Gene Symbol: PREP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112804.5 | NP_188548.2 | 5 - 6 | 1134 | 149 | |
| GenBank mRNA | AY090240.1 | - | 5 - 6 | 910 | 149 | |
| AY091051.1 | - | 5 - 6 | 908 | 149 | ||
| BT002372.1 | - | 5 - 6 | 844 | 149 | ||
| BT006362.1 | - | 5 - 6 | 844 | 149 | ||
More Information
Set Membership: |
|

PREP1
Genomic Map