Assay Details
| Gene Symbol: PARC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035648.1 | NP_001030725.1 | 8 - 9 | 1501 | 92 | |
| NM_112805.5 | NP_188549.2 | 8 - 9 | 1540 | 92 | ||
More Information
Set Membership: |
|

PARC6
Genomic Map