Assay Details
| Gene Symbol: ATG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338385.1 | NP_001327751.1 | 1 - 2 | 448 | 125 | |
| NM_001338386.1 | NP_001327752.1 | 1 - 2 | 448 | 125 | ||
| NM_112806.3 | NP_188550.3 | 1 - 2 | 376 | 125 | ||
| GenBank mRNA | HQ446214.1 | - | 1 - 2 | 208 | 125 | |
More Information
Set Membership: |
|

ATG2
Genomic Map