Assay Details
| Gene Symbol: ATCAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112832.4 | NP_188576.1 | 2 - 3 | 686 | 145 | |
| GenBank mRNA | AF370261.1 | - | 2 - 3 | 300 | 145 | |
| AY063076.1 | - | 2 - 3 | 204 | 145 | ||
| AY302081.1 | - | 2 - 3 | 204 | 145 | ||
More Information
Set Membership: |
|

ATCAD4
Genomic Map