Assay Details
| Gene Symbol: PUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112845.4 | NP_188589.1 | 2 - 3 | 1274 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUT5| Gene Symbol: PUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112845.4 | NP_188589.1 | 2 - 3 | 1274 | 84 | |
Set Membership: |
|