Assay Details
| Gene Symbol: TOM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338439.1 | NP_001326561.1 | 6 - 7 | 943 | 73 | |
| NM_112890.5 | NP_188634.1 | 6 - 7 | 751 | 73 | ||
| GenBank mRNA | AY050925.1 | - | 6 - 7 | 593 | 73 | |
| AY117216.1 | - | 6 - 7 | 528 | 73 | ||
More Information
Set Membership: |
|

TOM40
Genomic Map