Assay Details
| Gene Symbol: TOM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338439.1 | NP_001326561.1 | 9 - 10 | 1193 | 119 | |
| NM_112890.5 | NP_188634.1 | 9 - 10 | 1001 | 119 | ||
| GenBank mRNA | AY050925.1 | - | 9 - 10 | 843 | 119 | |
| AY117216.1 | - | 9 - 10 | 778 | 119 | ||
More Information
Set Membership: |
|

TOM40
Genomic Map