Assay Details
| Gene Symbol: UBC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112897.4 | NP_566653.1 | 1 - 2 | 350 | 62 | |
| GenBank mRNA | AY127573.1 | - | 1 - 2 | 239 | 62 | |
| BT025536.1 | - | 1 - 2 | 125 | 62 | ||
| DQ027033.1 | - | 1 - 2 | 125 | 62 | ||
More Information
Set Membership: |
|

UBC19
Genomic Map