Assay Details
| Gene Symbol: UBC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112897.4 | NP_566653.1 | 2 - 3 | 372 | 81 | |
| GenBank mRNA | AY127573.1 | - | 2 - 3 | 261 | 81 | |
| BT025536.1 | - | 2 - 3 | 147 | 81 | ||
| DQ027033.1 | - | 2 - 3 | 147 | 81 | ||
More Information
Set Membership: |
|

UBC19
Genomic Map