Assay Details
| Gene Symbol: PUM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338465.1 | NP_001319600.1 | 9 - 10 | 2797 | 71 | |
| NM_001338466.1 | NP_001327305.1 | 8 - 9 | 2905 | 71 | ||
| NM_001338467.1 | NP_001327304.1 | 8 - 9 | 2981 | 71 | ||
More Information
Set Membership: |
|

PUM5
Genomic Map